Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE8B Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
Phosphodiesterase 8B, is a high affinity, cAMP-specific phosphodiesterase. Enhanced PDE8B activity has been associated with the phenotypic expression of Gsalpha mutations in human pituitary tumors. PDE8B is absent in normal pituitary but present in GH-secreting adenomas. PDE8B activity is inhibited by dipyridamole, but it is insensitive to rolipram, zaprinast, or IBMX.

PDE8B antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-317
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PDE8B
Antigen name Phosphodiesterase 8B
Reactivity Human, Gorilla, Gibbon, Monkey
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms CRF2, CRF-RB, HM-CRF, CRFR2
Immunogen PDE8B antibody was raised against a peptide located near the internal domain of PDE8B (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty