Phosphodiesterase 7B, a cAMP-specific phosphodiesterase, has been shown to affect cAMP-mediated neural activity and cAMP metabolism in the brain. PDE7B activity is inhibited by dipyridamole, IBMX, and SCH51866. At least three splice variants have been isolated for PDE7B from rats and they show tissue specific expression patterns.
PDE7B antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PDE7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human