Anti-PAK6 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-308EA 462 GBP
PRSI48-308
Anti-PAK6 Rabbit Polyclonal Antibody
PAK6, a PAK (STE20) type protein kinase, contains a N-terminal Cdc42/Rac interactive binding (CRIB) motif and a C-terminal kinase domain. The protein has been shown to bind to GTP-Cdc42 and GTP-Rac; however, in contrast to most PAKs, kinase activity was not stimulated by Cdc42 or Rac, but could be stimulated by androgen receptor (AR) binding. PAK6 protein also has been shown to bind to estrogen receptor alpha.

PAK6 antibody can be used in immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PAK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Dog, Gibbon, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR