Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADGRB1 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
BAI1 is an Orphan-B GPCR with an unknown ligand. BAI1 is postulated to be a member of the secretin receptor family, an inhibitor of angiogenesis, and a growth suppressor of glioblastoma. Along with BAI1, there are two other brain-specific angiogenesis inhibitor genes, designated BAI2 and BAI3, which have similar tissue specificities and structures. However, only BAI1 is transcriptionally regulated by p53. Loss of BAI1 expression has been implicated in glioblastoma.

BAI1 antibody can be used in immunohistochemistry starting at 10 - 15 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Gorilla, Human, Monkey




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-300
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ADGRB1
Antigen name Adhesion G Protein-Coupled Receptor B1
Reactivity Gibbon, Human, Gorilla, Bovine, Monkey
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms KIAA4089, R75078, GDAIF, BAI1, Brain-Specific Angiogenesis Inhibitor 1
Immunogen BAI1 antibody was raised against a peptide located in the N-Terminal domain of BAI1 (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty