|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | FZD5 |
Antigen name | Frizzled 5 |
Reactivity | Human, Bovine, Dog, Pig, Gibbon, Gorilla, Monkey, Rabbit |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Rabbit |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen synonyms | hFz5, Hfz5, C2orf31, Fz-5, MGC129692, FzE5, HFZ5, DKFZP434E2135 |
Immunogen | Frizzled 5 antibody was raised against a peptide located in the N-Terminal extracellular domain of Frizzled 5 (Human). |
Form | Liquid |
Tested applications | Immunoaffinity Chromatography |
WRITE A REVIEW
REVIEWS (0)
Sort all by: