GPR1 is an Orphan-A GPCR with an unknown ligand. GPR1 was identified in a search for genes responsible for mediating brain reward and may have a role in drug addiction. It has also been shown to act as an HIV coreceptor in kidney mesangial cells (Tokizawa et al., 2000).
GPR1 antibody can be used in immunohistochemistry starting at 8.6 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human