Anti-P2RY8 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-245EA 462 GBP
PRSI48-245
Anti-P2RY8 Rabbit Polyclonal Antibody
P2RY8 is an Orphan-A GPCR with an unknown ligand. This protein shares low similarity to the Xenopus P2Y8 receptor (CAA68213.1, 30% protein similarity).

P2RY8 antibody can be used in immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: P2RY8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR