Anti-VIPR1 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-225EA 590 GBP
PRSI48-225
Anti-VIPR1 Rabbit Polyclonal Antibody
Antibodies
Vasoactive Intestinal Polypeptide Receptor 1 (VIPR1) is a member of the Vasoactive Intestinal Polypeptide subfamily. Vasoactive intestinal peptide (VIP) plays multiple roles in the nervous, endocrine, and immune systems as a neurotransmitter, a hormone, and a cytokine. VIPR1, a receptor for VIP, has been shown to expressed widely in normal tissues and is the predominant receptor subtype for VIP in cancers. VIP and its receptors, VIPR1 and VIPR2, are highly expressed in the immune system and modulate diverse T cell functions. Recently VIP-R1 has been shown to be a novel and potent facilitator of HIV-1 infection. Two isoforms are produced by alternative splicing.

VimmunoprecipitationR1 antibody can be used in immunohistochemistry starting at 14 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR