Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-223EA 462 GBP
PRSI48-223
Anti-MAPK3 Rabbit Polyclonal Antibody
ERK1, a MAPK-type protein kinase, is a member of the Ras-MAPK pathway. This pathway modulates the activity of transcription factors, structural proteins, and other protein kinases, leading to biological responses such as cell proliferation and differentiation. Inactive ERK1 associates with MAPK/ERK kinases (MEK) 1 and 2 in the cytosol. Upon stimulation, MEK1/2 phosphorylates and activates ERK1 causing dissociation of the MEK/ERK complex. A portion of the activated ERK1 pool forms dimers and translocates to the nucleus to activate nuclear proteins such as ELK-1 and MSK1. Cytosolic ERK 1 acts on a different set of substrates, including MNK1/2 and MAPKAP-K1.

MAPK3 antibody can be used in immunohistochemistry starting at 1:50, and immunofluorescence.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR