Anti-DDR1 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-219EA 603 GBP
PRSI48-219
Anti-DDR1 Rabbit Polyclonal Antibody
Antibodies
DDR1 is a DDR/TKT type protein kinase. DDR1 is activated by various types of collagen, including types I through IV. Binding of collagen to DDR1 protein results in autophosphorylation and a delayed but sustained tyrosine kinase activation. DDR1 may function in cell-to-cell interaction or recognition. At least three mRNA variants, resulting in different protein isoforms of 875, 913 and 919 amino acids, have been reported. DDR1 protein has been shown to be over expressed in human breast, ovarian, esophageal and pediatric brain tumors.

DDR1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR