Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-SPHK1 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
Sphingosine kinase (SPHK) phosphorylates sphingosine to form a bioactive lipid mediate, sphingosine 1-phosphate (S1P or SPP). S1P is a novel lipid messenger with both intracellular and extracellular functions. Intracellularly, it regulates proliferation and survival while extracellularly it is a ligand for EDG 1. Expression of SPHK1 and subsequent production of S1P promotes the G1-S transition, protects cells from apoptosis, stimulates migration and angiogenesis, and induces tumor formation in mice.

SPHK1 antibody can be used in immunohistochemistry starting at 40 μg/mL, and flow cytometry.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Antigen: SPHK1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Reactivity: Gibbon, Human, Monkey

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-207
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol SPHK1
Antigen name Sphingosine Kinase 1
Reactivity Gibbon, Monkey, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms SPHK 1, SPK 1, SPK1., SK1, sphingosine kinase 1, SK 1, SPHK, SPK
Immunogen SPHK1 antibody was raised against a peptide located in the N-Terminal domain of SPHK1 (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty