Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
ERK1, a MAPK-type protein kinase, is a member of the Ras-MAPK pathway. This pathway modulates the activity of transcription factors, structural proteins, and other protein kinases, leading to biological responses such as cell proliferation and differentiation. Inactive ERK1 associates with MAPK/ERK kinases (MEK) 1 and 2 in the cytosol. Upon stimulation, MEK1/2 phosphorylates and activates ERK1 causing dissociation of the MEK/ERK complex. A portion of the activated ERK1 pool forms dimers and translocates to the nucleus to activate nuclear proteins such as ELK-1 and MSK1. Cytosolic ERK 1 acts on a different set of substrates, including MNK1/2 and MAPKAP-K1.

MAPK3 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-188
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MAPK3
Antigen name Mitogen-Activated Protein Kinase 3
Reactivity Gibbon, Human, Monkey, Gorilla
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms ERK-1, HS44KDAP, ERK1, P44ERK1, p44-MAPK, P44MAPK, p44-ERK1, PRKM3, HUMKER1A, ERT2
Immunogen MAPK3 antibody was raised against a peptide located near the internal domain of MAPK3 (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty