Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRM Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
PTP mu, a R2A receptor protein tyrosine phosphatase, is composed of an extracellular segment containing a MAM domain, an immunoglobulin domain and four fibronectin type III repeats, a transmembrane segment, and two intracellular protein phosphatase domains. PTP mu has been shown to affect cell-cell aggregation, regulate cardiac myocyte Kv1.5 channel expression, and promote neurite outgrowth of retinal ganglion cells. The human PTP mu gene is localized on chromosome 18pter-q11, a region with frequent abnormalities implicated in human cancer. PTP mu may be important in responses to pathological conditions associated with the loss of cell-cell interactions in the heart.

PTPRM antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 - 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTPRM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Hamster, Horse, Human, Mouse, Rabbit, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-181
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PTPRM
Antigen name Protein Tyrosine Phosphatase Receptor Type M
Reactivity Gibbon, Rabbit, Horse, Rat, Mouse, Human, Hamster
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms R-PTP-MU, RPTPU, PTPRL1, MGC166994, RPTPM, hR-PTPu
Immunogen PTPRM antibody was raised against a peptide located near the internal domain of PTPRM (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty