Anti-ERBB4 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-178EA 462 GBP
PRSI48-178
Anti-ERBB4 Rabbit Polyclonal Antibody
ERBB-4 receptor tyrosine kinase is activated by neuregulins, nrg- 2, nrg-3, heparin-binding egf-like growth factor, betacellulin and ntak to regulate cell proliferation and differentiation. Several isoforms of the ERBB-4 receptor tyrosine kinase have been identified and differ functionally in their response to phorbol ester: they represent cleavable and noncleavable forms of the receptor. These isoforms are differentially expressed in tissues.

ERBB4 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 - 10 μg/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ERBB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR