Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NR4A1 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
Nak1, a NR4 Nerve Growth Factor-Like receptor, has been shown to affect the hypothalamic-pituitary axis, neuronal differentiation, neuroendocrine regulation of adrenocortical function, and T-cell apoptosis. Nak1 also is known as nuclear receptor Tr3 and is similar to mouse Nur77 and rat NGFI-B. It is encoded by one of the immediate early genes (IEG) in response to nerve growth factor and other serum growth factors. Nak1 can bind to DNA as a monomer, homodimer, or heterodimer (with retinoid X receptors). Nak1 has been shown to inhibit vascular lesion formation and could serve as a novel target in the treatment of vascular diseases.

NR4A1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NR4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Horse, Human, Monkey, Orangutan




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-174
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol NR4A1
Antigen name Nuclear Receptor Subfamily 4, Group A, Member 1
Reactivity Horse, Orangutan, Human, Gibbon, Monkey
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms Orphan nuclear receptor HMR, NAK1, Early response protein NAK1, HMR, TR3 orphan receptor, ST-59, Orphan nuclear receptor NR4A1, GFRP1, NR4A1
Immunogen NR4A1 antibody was raised against a peptide located near the internal domain of NR4A1 (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping