TAAR9/TA3 is an Orphan-A GPCR because the ligand has not been identified. TAAR9/TA3 was identified by degenerate PCR because of its similarity to other trace amine receptors. The close proximity on chromosome 6 and high sequence homology to the other trace amine receptors suggest that TAAR9/TA3 is likely to bind a related or unidentified amine.
TAAR9 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TAAR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human