BRS3 is classified as an Orphan-A Receptor and is highly similar to the bombesin receptors but has a very low affinity for bombesin itself. Mice lacking BRS3 develop metabolic defects and obesity, suggesting that BRS3 may be an important target for obesity research. In addition, BRS3 may be involved in diabetes and hypertension.
BRS3 antibody can be used in Western Blot starting at 0.5 μg/mL, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: BRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Guinea pig, Gorilla, Hamster, Horse, Human, Mouse, Rat