Anti-GPR124 Rabbit Polyclonal Antibody

Supplier: ProSci
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PRSI48-151EA 462 GBP
PRSI48-151
Anti-GPR124 Rabbit Polyclonal Antibody
Tumor endothelial marker 5 is an Orphan-U GPCR with an unknown ligand. Tumor Endothelial Marker 5, like GPR125, has a leucine rich repeat (LRR), an immunoglobulin (Ig) domain, and a hormone-binding domain (HBD). The Ig domain shows similarities to motilin and titin, while the LRR domain shows similarities to LRIG1 and SLIT1-2.

GPR124 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR124
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR