Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-TAAR1 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
TAAR1/TA1 is a Trace Amine Receptor activated by tyramine and beta-PEA. TAAR1/TA1 is likely to be involved in regulation of blood pressure, electrolyte homeostasis, and in controlling mood. An allelic variant of TAAR1/TA1 has been identified.

TAAR1 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Antigen: TAAR1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Reactivity: Chimpanzee, Human, Monkey

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-144
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol TAAR1
Antigen name Trace amine-associated Receptor 1
Reactivity Monkey, Human, Chimpanzee
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms TaR-1, TAR1, TRAR1, Trace amine receptor 1, TA1
Immunogen TAAR1 antibody was raised against a peptide located in the 3rd cytoplasmic domain of TAAR1 (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty