United Kingdom
Order Entry
Anti-LXN Goat Polyclonal Antibody
  PRSI45-825
undefined
Anti-LXN Goat Polyclonal Antibody
  PRSI45-825
 
£604.00
   
  1 * 100 µG

 

  • Antigen name:
    Latexin
  • Antigen symbol:
    LXN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    Endogenous carboxypeptidase inhibitor|ECI|Latexin protein|TCI|MUM|Tissue carboxypeptidase inhibitor.|Latexin
  • Concentration:
    500 ug/mL
  • Immunogen:
    Latexin antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of Latexin.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Latexin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Latexin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat