Infant/adult nutritional formula, NIST-1849A standard
Supplier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROONIST-1849AEA 2000 GBP
PROONIST-1849A
Infant/adult nutritional formula, NIST-1849A standard
This reference material is intended primarily for validation of methods for determining proximates, fatty acids, cholesterol, vitamins, elements, amino acids and nucleotides in infant and adult nutritional formulas and similar materials. It can be also used for the quality assurance to in-house reference materials.
These milk-based, hybrid infant/adult nutritional powder prepared by using infant formula and adult nutritional products. A unit consists of 10 packets, each containing approximately 10 g of material.
Certifications: Traceable to NIST and SRM.