Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Nucleic Acid Purification Components

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Hi-Bind® DNA mini columns are silica glass fibre columns that can bind up to 100 μg of genomic DNA or 35 μg of plasmid DNA per prep. Columns feature a...
Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, peqGOLD, SPW/CG Wash Buffer
Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, PerfectBind, PerfectBind RNA columns (Classic Line), red, 200/pack
Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, PerfectBind, PerfectBind DNA columns (Classic Line), violet, for peqGOLD Plasmid Miniprep Kit II, 200/pack

Supplier:  VWR Peqlab
Description:   PEQGOLD PERFECTBIND BLOOD DNA COLUMNS 200 pieces, Safety Line peqGOLD PerfectBind Blood DNA Columns (red) Shipment: RT Storage: RT 1 * 200 items
UOM:  1 * 200 items
Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, PerfectBind, PerfectBind Gelex columns (Classic Line), 200/pack

Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, peqGOLD, Solution I
UOM:  1 * 250 ml

Supplier:  VWR Peqlab
Description:   Nucleic Acid Purification, Components, PerfectBind, PerfectBind MicroSpin RNA columns
UOM:  1 * 200 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 8  of 12
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping