Disinfectant surface wipes, CHEMGENE HLD4H
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NORMXTM351EA 110 GBP
NORMXTM351
Disinfectant surface wipes, CHEMGENE HLD4H
These non woven wipes made from PP viscose are impregnated with a multi-active disinfectant solution that is alcohol free.
  • High strength and absorbency
  • Low lint properties
  • No requirement for pre-mixing , reducing wastage and risk errors
  • Canister can be stored on a work top or in a wall mounted bracket
CHEMGENE HLD4H high level disinfectant wipes are intended for cleaning and disinfecting of hard surfaces of non invasive, non lumened medical devices, such as monitors, IV poles and X-Ray equipment. Not to be used to disinfect invasive medical devices.
Bactericidal activity:
Acinetobacter calcoaceticus, Enterococcus hirae, Escherichia coli, Legionella pneumophila, listeria monocytogenes, Methicillin-resistant Staphylococcus aureus, Methicillin-resistant Staphylococcus pseudintermedius, Pseudomonas aeruginosa, Salmonella tryphimurium and Staphylococcus aureus.
Virucidal activity:
Adenovirus, Hepatitis B, Herpes Simplex, Human Immunodeficiency Virus (HIV) and Norovirus.
Mycobactericidal activity:
Bacillus cereus, Bacillus subtilis, Mycobacterium avium, Mycobacterium bovis and Mycobacterium terrae.
Fungicidal activity:
Aspergillus niger, Candida albicans and Trichophyton rubrum
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904