Water, Baxter® for irrigation (WIR), sterile
Supplier: DISTRIBUTOR UK
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DIUK3KB7114EA
128
GBP
DIUK3KB7114
Water, Baxter® for irrigation (WIR), sterile
Water
Sterile water for irrigation.
Packaging: In a Viaflex container.
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...