Copper(II) sulphate pentahydrate 98.5-100.5% (dried basis), granular USP, Macron Fine Chemicals™
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Copper sulphate pentahydrate
, Cupric sulphate pentahydrate
4752-12EA
0
GBP
4752-12
MACR4752-10
Copper(II) sulphate pentahydrate 98.5-100.5% (dried basis), granular USP, Macron Fine Chemicals™
Copper (II) sulphate pentahydrate
- GMP manufactured product
Formula:
CuSO₄·5H₂O MW: 249.69 g/mol Density: 2.28 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3288 ADR: 6.1,III REACH: 01-2119520566-40 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Preserve in tight containers. Store at 25°C, excursions permitted 15-30°C. | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (CuSO₄·5H₂O) (dried basis) | 98.5 - 100.5 % |
USP - Calcium (Ca) | ≤0.005 % |
Solubility (5 in 50) | Passes Test |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Iron (Fe) | ≤0.003 % |
USP - Nickel (Ni) | ≤0.005 % |
USP - Loss on Drying | 33.0 - 36.5 % |
USP - Potassium (K) | ≤0.01 % |
USP - Sodium (Na) | ≤0.02 % |
Appearance (blue, crystalline granules) | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...