FastDNA-96™ soil microbe DNA kit
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FastDNA™
ICNA119696200EA 1220 GBP
ICNA119696200
FastDNA-96™ soil microbe DNA kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastDNA-96™ Soil Microbe DNA Kit quickly and efficiently isolates humic-free, PCR-ready genomic DNA from soil samples in approximately 50 minutes.
Gram(+/-) bacteria, fungi, plant and animal tissues, algae, spores and many other members of a soil population, including clay, sandy, silty, peaty, chalky, and loamy soils, are efficiently lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
Delivery information: FastDNA-96™ lysing matrix rack (0,5 mm beads) (2×96-well rack); lysis buffer (2×40 ml); soil DNA binding solution (150 ml); binding plate pre-wash buffer (50 ml); soil DNA wash buffer (100 ml); elution solution (2×10 ml); inhibitor plate prep solution (30 ml); deep-well plate (2 each); MP-96 binding plate (2 each); MP-96 inhibitor removal plate (2 each); collection plate (2 each); elution plate (6 each); foil plate cover (4 each).