Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Couch covers, SCOTT® ESSENTIAL

Supplier: Kimberly-Clark
SCOTT® ESSENTIAL
This single ply couch cover is strong and resistant.

  • For use in busy areas
  • Can be used with a dispensing system to help control use and reduce wastage

Ideal for: everyday treatment couches protection whilst maintaining required levels of hygiene in the healthcare environment.




 

ORDER

Expand All / Collapse All
Description L×W Colour Packed Pk VWR Catalog Number Quantity
SCOTT® ESSENTIAL couch cover roll, 1 ply 510×380 mm Blue 12 rolls of 140 sheets 12 Roll 115-2011

Dispenser
Description Pk VWR Catalog Number Quantity
Wall-mountable roll dispenser, 7056, white, for rolls with max. 500 mm width 1 115-2180
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping