UV/Vis spectrophotometer, Libra S60PC
Supplier: Biochrom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Libra
WAPR80-7000-10EA 0 GBP
WAPR80-7000-10
UV/Vis spectrophotometer, Libra S60PC
Spectrophotometers and Spectrometers UV/Visible Spectrophotometers
Libra S60PC is designed to deliver reliable UV/Vis spectrophotometry in multiple users environments especially in life science.
  • Full PC control with resolution software
  • Built-in full suite of software applications with data export methods
  • Colour touch screen user interface with menu driven software and intuitive icons
  • Trace Manager enables the user to manipulate data on the instrument or USB stick
  • Multi user log allows supervisors to restrict access to software functionality allowing for secure methods
Applications include DNA, RNA and Oligo concentration and purity measurement with wavelength scanning, CyDye™ DNA quantitation, Tm calculation, UV protein concentration measurement, Bradford, Biuret, Lowry or BCA colorimetry.