Anti-EMSY Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-13069R-A647
: Bioss
- Antibody type:Primary
- Antigen symbol:EMSY
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
EMSY binds BRCA2 within a region (exon 3) deleted in cancer. EMSY is capable of silencing the activation potential of BRCA2 exon 3, associates with chromatin regulators HP1 and BS69, and localizes to sites of repair following DNA damage. Its levels are amplified in breast and ovarian cancer.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EMSY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat