Water standards for Karl Fischer titration, Aquametric
Supplier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APPC395459.2527EA 101 GBP
APPC395459.2527
Water standards for Karl Fischer titration, Aquametric
Moisture standards to determine the titre of the Aquametric Karl Fischer reagents.
Certifications: Reference substances are traceable against SRM of NIST.