Dual adjustable nucleic acid sequencing tanks

Supplier: CBS Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

730-0013EA 2380 GBP
CBSCDASG-600-33 CBSCDASG-500-20 CBSCDASG-500-33 CBSCDASG-600-20 CBSCDASG-400-20 CBSCDASG-400-33
Dual adjustable nucleic acid sequencing tanks
The nucleic acid sequencers are adjustable to accommodate plates with path lengths from 22 cm up to the maximum length of the unit, providing the versatility to run short gels for oligonucleotide separations as well as preparative DNA separations or long gels for analytical sequencing on the same unit. Applications include differential display, DNA fingerprinting, DNA footprinting, DNA sequencing, genotyping, mutation detection, micro-satellites/SSTR, mini-satellites, RNase protection, and SDS-PAGE.

  • Available in 200 mm or 330 mm widths, with a height capability of 420 mm, 520 mm or 620 mm
  • Upper reservoir is divided in half, allowing the user to run identical or independent buffer conditions on either side
  • Rotating base allows the unit to swivel, providing easy access to both sides
  • Both upper and lower reservoirs have drains with tubing adapters and safety covers with attached leads
  • Reservoirs are easily detached for disposal of radioactive buffer solutions or routine cleaning

Ordering information: Aluminium plates and bar clamps must be ordered separately.

Delivery information: Each sequencer comes equipped with rotating base, divided upper reservoir, two sets of 62 well sharktooth combs (0,4 mm), two sets of glass plates (420 mm), two spacer sets 0,4×420 mm (h), one roll of gel sealing tape, two wedge plate separators, two gel thermometer strips, bottle of Gel Repel (29,5 ml) and 12× white spring clamps.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR