United Kingdom
Order Entry
Anti-FZD6 Rabbit Polyclonal Antibody
Anti-FZD6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13218R
Supplier:  Bioss
Anti-FZD6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13218R
Supplier:  Bioss
Price
£680.00
Log in for availability
Unit of Measure 1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Frizzled 6
  • Antigen symbol:
    FZD6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    8323
  • Antigen synonyms:
    HFZ6|FZ-6|NDNC10|FZ6
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    19-50/706
  • Purification:
    Purified by Protein A
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: FZD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat