Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Anti-Mouse IgG2a Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: MP Biomedicals

This antibody is specific for the gamma 2a chain. It exhibits minimal to no cross reactivity to other mouse IgG subclasses, IgA,IgM, or light chains. There is minimal or no cross reaction with human serum proteins.

  • Working dilution: 1:1000 - 1:2000 for ELISA
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horse radish peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µG ICNA08642541
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG2a
Antigen name Immunoglobulin G2a
Reactivity Mouse
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody type Secondary
Host Rabbit
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Purification Affinity purified
Form Liquid
Storage temperature 0 to +5 °C
Tested Applications Immunoblot
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty