Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti IgE Human Polyclonal Antibody

Supplier: MP Biomedicals
By Immunoelectrophoresis, single arc was observed against antisera to whole human serum, human IgE and human kappa light chain.

  • Supplied in Tris or phosphate buffered saline

It is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

It is prepared from plasma shown to be nonreactive for HBsAg, anti-HCV, anti-HBc and negative for anti-HIV 1 and 2 by FDA-required tests.

Type:
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Myeloma source
Isotype:
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG ICNA08643491
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgE
Antigen name Immunoglobulin E
Reactivity Human
Clonality Polyclonal
Host Myeloma source
Storage temperature 0...5 °C
Tested applications Immunoelectrophoresis

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty