Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Mouse IgG2b Antibody

Supplier: MP Biomedicals
This antibody is prepared by the mixture of immunoglobulins from two different tumor sources such as MOPC 195 and MOPC 141, which provides a boader spectrum of antigenic sites associated with the normal immunoglobulin class or subclass.

Based on gel diffusion and IEP analysis, it contains only IgG2B myeloma proteins.

Type:
Antigen: IgG2b
Clonality:
Clone:
Conjugation:
Epitope:
Host: Myeloma source
Isotype:
Reactivity: Mouse



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 mg 1 mg ICNA08643371
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG2b
Antigen name Immunoglobulin G2b
Reactivity Mouse
Host Myeloma source
Concentration 1,0 mg/ml
Purification Affinity purified
Form Liquid
Storage temperature −80 °C
Purity 95%
Tested applications Immunoelectrophoresis, Gel Diffusion

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping