Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Anti-Human IgG1 Monoclonal Antibody (Biotin)

Supplier: MP Biomedicals
These antibodies specifically reacts with human IgG1 (Fc region).

  • Working dilution: 1:500 - 1:1000 for most applications

Type: Secondary
Antigen: IgG1 Fc
Clonality: Monoclonal
Clone: HP6069
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1k
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
500 μL 500 µl ICNA08634721
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG1
Antigen name Immunoglobulin G1
Reactivity Human
Conjugation Biotin
Clonality Monoclonal
Antibody type Secondary
Clone HP6069
Host Mouse
Isotype IgG1k
ImmunoChemistry Yes
Form Lyophilized
Antibody fragment F(c)
Storage temperature 0 to +5 °C
Tested applications Immunoblot

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty