Mouse Anti-Human IgE Monoclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ICNA08634361EA 0 GBP
ICNA08634361
Mouse Anti-Human IgE Monoclonal Antibody
These antibodies specifically reacts with human IgE heavy chain (Fc region).

Type: Secondary
Antigen: IgE Fc
Clonality: Monoclonal
Clone: HP6061
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR