Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Anti-Human IgE Monoclonal Antibody

Supplier: MP Biomedicals
This purified monoclonal antibody is supplied in 10 mM phosphate buffered saline, pH 7,4.

  • Dilution range: 10 µg/ml in PBS for EIA
  • Specificity: Human IgE heavy chain (Fc region)

Type: Secondary
Antigen: IgE Fc
Clonality: Monoclonal
Clone: HP6029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2ak
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
200 μg 200 µG ICNA08634371
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgE
Antigen name Immunoglobulin E
Reactivity Human
Clonality Monoclonal
Antibody type Secondary
Clone HP6029
Host Mouse
Isotype IgG2ak
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Concentration 2,0 mg/ml
Form Liquid
Antibody fragment F(c)
Storage temperature +2 °C to +8 °C
Tested applications Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty