Cottonseed oil, liquid

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ICNA0290141901EA 81.1 GBP
ICNA0290141901
Cottonseed oil, liquid
Cottonseed oil

  • Iodine Value: 109 - 120
  • Saponification Number: 190-198 (Lit.)


Soluble in 400 mg plus 4 ml of chloroform - clear, faint yellow to yellow-green.

Density: 0.92 g/ml (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00130872
CAS Number: 8001-29-4

Order Now

Specification Test Results

Identity Test Passes
Source Gossypium hirsutum
Myristic (C-14:0) 0,5 - 2,0%
Palmitic (C-16:0)% 17 - 29
Steric (C-18:0) 1,0 - 4,0%
Oleic (C-18:1) 13 - 44%
Linoleic (C-18:2) 40 - 63%
Linolenic (C-18:3) 0,1 - 2,1%
Iodine Value 109 - 120
Specific Gravity 0,915 - 0,921

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR