Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LDS sample loading buffer, non-reducing (4X)

Supplier: VWR Chemicals
LDS Sample Loading Buffer, Non-reducing (4X) is used to prepare protein samples for denaturing polyacrylamide gel electrophoresis using Bis-Tris and Laemmli gels. Mixing LDS Sample Loading Buffer with protein samples only denatures the proteins without reducing them.

  • Premixed for convenience
  • Requires cold storage
  • 4X concentration allows more volume for protein per lane
  • Bring to room temperature prior to use

Reducing agents such as DTT or 2-mercaptoethanol can be added to the buffer for protein sample preparation under reducing conditions. Glycerol is included in the loading buffer as a density agent to ensure the samples sink into the sample wells, while Coomassie serves as a tracking dye to monitor the progression of electrophoresis.

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
LDS sample loading buffer, non-reducing (4X), 5 ml 1 1B1669-5ML
LDS sample loading buffer, non-reducing (4X), 25 ml 1 1B1669-25ML
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty