Would you like to log in again?
Maintenance kit for electrodes, complete, GK ANNEX (Radiometer Analytical)
Supplier: Hach
Hach®
The GK ANNEX Maintenance Kit contains all that is needed for regular maintenance of combined pH, glass, and reference electrodes with saturated potassium chloride salt bridge.
Kit contains: solutions for normal and intensive cleaning, KCl as saturated solution and crystals for refilling.
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
GK ANNEX electrode maintenance kit, complete 1 KIT HACHS91M001
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty