3D gloss meter
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
664-0223EA 4860 GBP
664-0223
3D gloss meter
Coating Testers
This multiangular reflectometer is designed with a three-angle geometry perfectly suited to each and every field of gloss measurement.
Suitable for metallic and non-metallic surfaces.
Certifications: ISO 7668