Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescent cell counter, Cellometer® K2

Supplier: Nexcelom Bioscience
Cellometer®
The Cellometer K2 cell counter is designed for automated brightfield and fluorescent image capture and analysis. It can perform cell count, size, viability, intensity and population analysis, and viability measurements.

  • Cell staining in heterogeneous samples
  • Cell count in clumps and irregular-shaped cells
  • Disposable counting chambers – No wash steps

Designed for cells lines and primary cells in complex samples: Heavy debris, red blood cell contamination, low cell concentrations, and multiple cell types, including PBMCs, stem cells, cancer cells, splenocytes, hepatocytes and whole blood. The K2 can perform cell-based assays such as apoptosis, cell cycle, and GFP analysis.



 

ORDER


The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Counting chambers with protective film 75 734-2886
Counting chambers with protective film 900 734-2887
Counting chambers with protective film 2.700 NEXCCHT4-SD100-314
Disposable plastic counting chambers 2.500 734-2885
Disposable plastic counting chambers 1.500 734-2884
Disposable plastic counting chambers 500 734-2883
Disposable plastic counting chambers 100 NEXCCHT4-PD100-002
FCS Express 4 software upgrade 1 NEXCSWUG-CBA-10
Laptop computer for Cellometer® 1 NEXCCMCOMPSK150
Validation solution for Cellometer® fluorescent instruments (100 tests) in a vial 2 ml NEXCCCBM-011-2ML
ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml) 1 NEXCCS2-0106-5ML
ViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml) 1 NEXCCS2-0107-5ML
ViaStain™ propidium iodide (PI) staining solution for dead nucleated cells 5 ml NEXCCS2-0108-5ML
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty