Atomic absorption spectrocopy (AAS) standards, phosphorus in oil, Certipur®, Supelco®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Certipur®
1.15072.0100EA 172 GBP
MERC1.15072.0100
Atomic absorption spectrocopy (AAS) standards, phosphorus in oil, Certipur®, Supelco®
Standard dissolved in oil is ready-to-use calibration standards for the analysis of phosphorus by AAS in a non-aqueous matrix.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022