4-(Carboxymethyl)phenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-(Carboxymethyl)phenylboronic acid Pinacol ester
, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid
, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
, 2-[(4-Carboxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
ACRO444670010EA
143
GBP
ACRO444670010
4-(Carboxymethyl)phenylboronic acid pinacol ester 97%
4-(Carboxymethyl)phenylboronic acid pinacol ester
Formula:
C₁₄H₁₉BO₄ MW: 262.1093 g/mol Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038755 CAS Number: 797755-07-8 |
Specification Test Results
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | 162 to 168 °C |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...