9-Methylcarbazole 99%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
9-Methyl-9H-carbazole
, N-Methylcarbazole
ACRO441050050EA
80
GBP
ACRO441050050
9-Methylcarbazole 99%
9-Methylcarbazole
Formula:
C₁₃H₁₁N MW: 181.2331 g/mol Boiling Pt: 163…167 °C (.4Torr) Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013431 CAS Number: 1484-12-4 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 87 to 92 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...