4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid 97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
ACRO438260050EA
173
GBP
ACRO438260050
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid 97%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Formula:
C₉H₁₀O₂S MW: 182.24 g/mol Melting Pt: 196.1…202.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464469 CAS Number: 40133-07-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 195 to 205 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...