2,4,6-Trimethoxybenzylamine hydrochloride 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
ACRO437330050EA
204
GBP
ACRO437330050
ACRO437330010
2,4,6-Trimethoxybenzylamine hydrochloride 98%
2,4,6-Trimethoxybenzylamine hydrochloride
Formula:
C₁₀H₁₆ClNO₃ Melting Pt: 204…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012855 CAS Number: 146548-59-6 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...