5-Cyano-3-pyridinylboronic acid 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
ACRO430400010EA
210
GBP
ACRO430400010
5-Cyano-3-pyridinylboronic acid 97%
5-Cyano-3-pyridinylboronic acid
Formula:
C6H5BN2O2 MW: 147.9300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07374882 CAS Number: 497147-93-0 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...