Ultra low temperature freezer, TSX series
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
REVCTSX600VEA 0 GBP
REVCTSX600V
Ultra low temperature freezer, TSX series
Thermo Scientific™ TSX Series feature V-drive technology and providing temperature uniformity.
  • Water-blown foam insulation
  • Natural refrigerants for lower environmental impact and higher cooling efficiency
  • With intuitive touch-screen user interface
These V-drive freezers runs at variable speeds, adjusting cooling performance to the conditions inside and outside the freezer. When conditions are stable, the V-drive runs at a low speed, reducing energy consumption while maintaining a stable temperature for sample protection. When there are frequent door openings, or samples are added to the freezer, the system detects the activity and increases the drive speed.